missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide solution, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis, Solstice

Catalog No. 60045826
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Glass Bottle
Plastic Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045826 6 x 1 L Plastic Bottle
6000320 1 L Plastic Bottle
6000322 2.5 L Plastic Bottle
6000324 5 L Plastic Bottle
60045827 6 x 1 L Glass Bottle
60045823 4 x 2.5 L Plastic Bottle
60045825 4 x 5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60045826 Supplier Solstice Supplier No. 305016X1L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

Boiling Point 24°C
CAS Min % ≥70.0000%, ≥20.0000%
CAS Max % <90.0000%, <30.0000%
Linear Formula NH4OH
UN Number UN2672
Beilstein 3587154
Density 0.892-0.910 g/mL at 20 °C; 0.9 g/mL at 25 °C(lit.)
Quantity 6 x 1 L
Percent Purity >25%
Grade puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Ammonium hydroxide solution, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis
Show More Show Less
ShelfLife 1440 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Ammonia solutions
Recommended Storage Room Temp