missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 50% (v/v) Solution, Spectrum™ Chemical

Catalog No. 18608691
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608691 500 mL Poly Bottle
18608683 1 L Poly Bottle
18608682 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608691 Supplier Spectrum Chemical Mfg Cor Supplier No. A160500MLPL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5

Specifications

CAS Max % 1
Quantity 500 mL
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Ammonium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.