missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide, 10% v/v aq. soln.

Catalog No. AA35575AP
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35575AP 500 mL
AA35575K2 1 L
AA35575K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35575AP Supplier Thermo Scientific Chemicals Supplier No. 035575AP

CAS: 1336-21-6 | H5NO | 35.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
SMILES N.O

Specifications

Color Colorless
Linear Formula NH4OH
UN Number UN2672
Concentration 10% v/v aq. soln.
Density 0.98 g/mL
IUPAC Name azanium;hydroxide
Formula Weight 35.05
Odor Ammonia-like
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Ammonium hydroxide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMMONIA SOLUTION
EINECSNumber 215-647-6
RTECSNumber BQ9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only