missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Glycyrrhizate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014538
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014538 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014538 Supplier Merck Emd Millipore Supplier No. PHR1689200MG
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Ammonium Glycyrrhizate is the ammonium salt of glycyrrhizic acid (GA), an important ingredient in the liquorice root with a broad-spectrum of biological and pharmacological properties. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 53956-04-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C42H62O16 · NH3
MDL Number MFCD00167400
Synonym Glycyrrhizic acid ammonium salt
Type Certified Reference Material
Quantity 200 mg
Formula Weight 839.96
Grade Certified Reference Material