missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, 98+%, ACS reagent

Catalog No. AC423385000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423385000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC423385000 Supplier Thermo Scientific Chemicals Supplier No. 423385000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190.0°C
Color White
pH 3.8 to 4.4 (5% soln. at 25°C)
Physical Form Crystalline Solid
Quantity 500 g
Assay Percent Range 98+%
Linear Formula NH4H2PO4
Merck Index 15, 541
Solubility Information Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
IUPAC Name azanium;dihydrogen phosphate
Formula Weight 115.03
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Density 1.8
Chemical Name or Material Ammonium dihydrogen phosphate
Show More Show Less
EINECSNumber 231-764-5
TSCA TSCA

RUO – Research Use Only