missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium cobalt(II) sulfate hexahydrate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼120°C (decomposition) |
| Quantity | 500 g |
| UN Number | UN3077 |
| Formula Weight | 395.21 (287.13 Anhydrous) |
| Percent Purity | 98% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 1.902 |
| Chemical Name or Material | Ammonium cobalt (II) sulfate hexahydrate |
Safety and Handling
| Hazard Category | H302-H317-H350 |
| Hazard Statement | GHS H Statement H302-H317 Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 237-043-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only