missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Citrate Dibasic, ACS, 98-103%, Spectrum™ Chemical

Catalog No. 18608625
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608625 500 g Poly Bottle
18608622 125 g Poly Bottle
18608621 2.5 kg Poly Bottle
18608623 12 kg Poly Bottle
18608624 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608625 Supplier Spectrum Chemical Mfg Cor Supplier No. A1175500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3012-65-5

Specifications

CAS Max % 1
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Molecular Weight (g/mol) 226.19
Percent Purity 98 to 103%
Grade Reagent
Packaging Poly Bottle
Residue after Ignition 0.0001
Chemical Name or Material Ammonium Citrate Dibasic

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.