missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Granular, Reagent, ACS, 99.5%, Spectrum™ Chemical

A1165, 12125-02-9, NH4Cl

Supplier:  Spectrum Chemical Mfg Cor A116545KGBL

 View more versions of this product

Catalog No. 18600593


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Ammonium Chloride, Granular, Reagent, ACS is an inorganic compound, that presents itself as a white crystalline salt that is highly soluble in water. It occurs from the reaction of hydrochloric acid and ammonia. It has uses in medicine as an expectorant in cough medicine, in metalwork as a flux in preparing metals to be tin coated as well as in laboratories to produce cooling baths at low temperatures. As an ACS grade Reagent, Spectrum Chemical manufactured Ammonium Chloride is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications

Specifications

12125-02-9
4.5-5.5
ClH4N
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
99.5%
Fiber Drum
100%
45 kg
MFCD00011420
N.Cl
53.49
ACS
0.01%

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
N.Cl
ClH4N
MFCD00011420
amine hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions