missing translation for 'onlineSavingsMsg'
Learn More

Ammonia-Ammonium Chloride Buffer TS, Ricca Chemical

Catalog No. 623516
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
623516 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 623516 Supplier Ricca Chemical Company Supplier No. 623.516
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

USP Formulation

Specifications

Chemical Name or Material Ammonia-Ammonium Chloride Buffer
CAS 7732-18-5, 1336-21-6, 12125-02-9
For Use With (Application) Compendial Testing
Physical Form Solution
Quantity 500 mL
Packaging Natural Poly Bottle