missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride-Ammonium Hydroxide, Buffer Solution, pH 10.0 ±0.1, For Water Hardness, APHA, Spectrum™ Chemical
Description
Spectrum™ Chemical Ammonium Chloride-Ammonium Hydroxide, Buffer Solution, pH 10.0 ± 0.1, For Water Hardness, APHA is an aqueous inorganic buffer solution concentrate that, when diluted, is used for the American Public Health Association's (APHA) method for determination of water hardness. Find the high quality buffer solution Ammonium Chloride-Ammonium Hydroxide at Spectrum. As an aqueous inorganic buffer solution concentrate, it is used for the American Public Health Association's (APHA) method for determination of water hardness after dilution.
Specifications
Specifications
| Chemical Name or Material | Ammonium Chloride-Ammonium Hydroxide |
| CAS | 1336-21-6, 12125-02-9, 7732-18-5 |
| CAS Max % | 1%, 98% |
| UN Number | UN2672 |
| Grade | APHA/EPA |
| Packaging | Amber Glass Bottle |
| pH | 10 ±0.1 |
| Quantity | 1 L |
| Physical Form | Chemical Solution |