missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(IV) Nitrate 98.0+%, TCI America™

Catalog No. C180650G
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C180650G 50 g
C1806500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C180650G Supplier TCI America Supplier No. C180650G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
MDL Number MFCD00151121
Synonym Ceric Ammonium Nitrate, CAN, Cerium(IV) Diammonium Nitrate, Diammonium Cerium(IV) Nitrate

Specifications

Color Red-Yellow
Physical Form Crystalline Powder
Quantity 50 g
UN Number 1477
Molecular Weight (g/mol) 548.22
Formula Weight 548.22
Percent Purity ≥98.0% (T)
Chemical Name or Material Ammonium Cerium(IV) Nitrate