missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium cerium(IV) nitrate, 99%, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
Specifications
Specifications
| Melting Point | 107.0°C to 108.0°C |
| Color | Colorless to Yellow |
| Physical Form | Fuming Liquid |
| Quantity | 250 g |
| Packaging | Glass bottle |
| Assay Percent Range | 99% |
| Density | 1.05 |
| Linear Formula | (NH4)2Ce(NO3)6 |
| Fieser | 01,120; 02,63; 03,44; 04,71; 05,101; 06,99; 09,99; 10,79; 15,70; 16,66; 17,68 |
| Merck Index | 15,1991 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. May be corrosive to metals. Harmful if swallowed. Causes severe skin burns and eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately |
| EINECSNumber | 240-827-6 |
| TSCA | TSCA |
RUO – Research Use Only