missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Buffer, LabChem™

Numéro de catalogue. LC109712
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
LC109712 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. LC109712 Fournisseur LabChem Code fournisseur LC109712
Il en reste null

Spécifications

Chemical Name or Material Ammonium Buffer
CAS 7732-18-5, 12125-02-9, 1336-21-6
Color Colorless
CAS Max % 89.1, 6.6, 4.3
Decomposition Information Gaseous Ammonia; Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Density 0.94g/mL
Physical Form Liquid
Quantity 1 L
Identification Passes Test
Linear Formula NH4Cl
Packaging Poly Bottle
UN Number UN2672
Specific Gravity 0.94g/mL
Solubility Information Soluble in water
Formula Weight 53.49
Afficher plus de résultats Afficher moins de résultats