missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate (Crystalline/HPLC), Fisher Chemical

Catalog No. A639500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A639500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A639500 Supplier Thermo Fisher Scientific Supplier No. FLA639500
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point 114°C
Color White
pH 7.0
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥97 %
Linear Formula CH3COONH4
Formula Weight 77.08
Percent Purity ≥97%
Grade Also meets ACS specifications
Identification Pass Test
Packaging Poly Bottle
Assay ≥97.0%
Chemical Name or Material Ammonium Acetate
Show More Show Less