missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate Buffer, For Iron, APHA 3500-Fe B, Certified, LabChem™
Supplier: LabChem LC109704
Specifications
| Ammonium Acetate Buffer | |
| 64-19-7 , 631-61-8 , 7732-18-5 | |
| 68.75,22.36,8.89 | |
| Liquid | |
| Passes Test | |
| CH3COONH4 | |
| UN2790 | |
| 77.08 |
| For Iron | |
| Colorless | |
| Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors; Gaseous Ammonia | |
| 4 L | |
| CH3COONH4 | |
| Poly Bottle | |
| Soluble in water | |
| Certified |