missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate ACS AR,Crystal, Macron Fine Chemicals™
Specifications
Specifications
| Quantity | 2.5kg |
| Formula Weight | 77.08 |
| Grade | ACS |
| Packaging | Poly |
| Chemical Name or Material | Ammonium Acetate |