missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, ACS, 97.0% min

Catalog No. AA1159936
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1159936 500 g
AA11599A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1159936 Supplier Thermo Scientific Chemicals Supplier No. 01159936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent in analytical chemistry, drugs | CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol

Ammonium acetate is a reagent used in chromatographic analysis of various compounds such as oligos, proteins, and peptides. It is also employed with acetic acid to provide a buffer solution. Generally it behaves as a catalyst in many reactions like Knoevenagel, Borch. It is also employed as a nutrient additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point ∽114°C (decomposition)
Physical Form Crystalline
Quantity 500 g
Flash Point 136°C (277°F)
Sensitivity Hygroscopic
Assay Percent Range 97.0% min
Density 1.17
Beilstein 4186741
Solubility Information Soluble in cold water,methanol ,SO2and acetone.
Formula Weight 77.08
Grade ACS
Chemical Name or Material Ammonium acetate
Show More Show Less
EINECSNumber 211-162-9
RTECSNumber AF3675000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only