missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate - Acetic Acid Solution, TAPPI for Iron, Ricca Chemical

Catalog No. 60216
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60216 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60216 Supplier Ricca Chemical Company Supplier No. RI60.216

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Ammonium Acetate - Acetic Acid Solution
CAS 7732-18-5, 64-19-7, 631-61-8
Color Colorless
CAS Min % 75.0, 15.71, 7.49
CAS Max % 78.0, 16.35, 7.79
Physical Form Liquid
Quantity 500 mL
Synonym dihydrogen oxide, dihydrogen monoxide
Grade Laboratory
Packaging Natural Poly Bottle