missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, ∽98%, MP Biomedicals
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 500 g |
| Assay Percent Range | ∽98% |
| Formula Weight | 77.1 |
| Percent Purity | 98% |
| Grade | Molecular Biology |
| Chemical Name or Material | Ammonium Acetate |
Safety and Handling
| Recommended Storage | Room Temperature |