missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ammonium Acetate, Ultra Pure Grade
Ammonium Acetate, Ultra Pure Grade is used in the purification and precipitation of DNA and for protein crystallization.
$45.05 - $209.16
Specifications
| Assay | ≥97% |
|---|---|
| Chemical Name or Material | Ammonium Acetate |
| CAS | 631-61-8 |
| Content And Storage | Room Temperature |
| Density | 1.17 at 68° F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19384890
|
MP Biomedicals™
0219384890 |
500 g |
Each for $150.39
|
|
|||||
|
ICN19384891
|
MP Biomedicals™
0219384891 |
1 kg |
Each for $209.16
|
|
|||||
|
ICN19384880
|
MP Biomedicals™
0219384880 |
100 g |
Each for $45.05
|
|
|||||
Description
- Ammonium acetate, ultra pure, is for applications which require tight control of elemental content.
- Ammonium acetate is a widely used reagent in molecular biology and chromatography.
- Its applications include the purification and precipitation of DNA and protein crystallization.
- Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides.
- Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.
Specifications
| ≥97% | |
| 631-61-8 | |
| 1.17 at 68° F | |
| 237.2° F | |
| Ultrapure | |
| Crystalline Powder |
| Ammonium Acetate | |
| Room Temperature | |
| 77.1 | |
| C2H7NO2 | |
| unii-rre756s6q2 |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.