missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ammonium Acetate, Ultra Pure Grade
Description
- Ammonium acetate, ultra pure, is for applications which require tight control of elemental content.
- Ammonium acetate is a widely used reagent in molecular biology and chromatography.
- Its applications include the purification and precipitation of DNA and protein crystallization.
- Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides.
- Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.
Specifications
Specifications
| Assay | ≥97% |
| Chemical Name or Material | Ammonium Acetate |
| CAS | 631-61-8 |
| Content And Storage | Room Temperature |
| Density | 1.17 at 68° F |
| Formula Weight | 77.1 |
| Melting Point | 237.2° F |
| Molecular Formula | C2H7NO2 |
| Purity | Ultrapure |
| Quantity | 500 g |
| Show More |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.