missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ammonium Acetate, Ultra Pure Grade

Ammonium Acetate, Ultra Pure Grade is used in the purification and precipitation of DNA and for protein crystallization.

Supplier:  MP Biomedicals™ 0219384880

 View more versions of this product

Catalog No. ICN19384880


Only null left
Explore available promotions

Description

Description

  • Ammonium acetate, ultra pure, is for applications which require tight control of elemental content.
  • Ammonium acetate is a widely used reagent in molecular biology and chromatography.
  • Its applications include the purification and precipitation of DNA and protein crystallization.
  • Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides.
  • Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.
Specifications

Specifications

≥97%
631-61-8
1.17 at 68° F
237.2° F
Ultrapure
unii-rre756s6q2
Ammonium Acetate
Room Temperature
77.1
C2H7NO2
100 g
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.