missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, 40% (w/w), Ricca Chemical
Chemical Identifiers
| CAS | 631-61-8 , 7732-18-5 |
|---|---|
| Molecular Formula | C2H7NO2 |
| Molecular Weight (g/mol) | 77.083 |
| InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
| Synonym | ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 |
| PubChem CID | 517165 |
| ChEBI | CHEBI:62947 |
| IUPAC Name | azanium;acetate |
| SMILES | CC(=O)[O-].[NH4+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
62016
|
N/A | 500 mL | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
62032
|
N/A | 1 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
6201
|
N/A | 4 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
R062000020F
|
N/A | 20 L | Cubitainer |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 631-61-8 , 7732-18-5 | |
| 77.083 | |
| ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
| CHEBI:62947 | |
| CC(=O)[O-].[NH4+] |
| C2H7NO2 | |
| USFZMSVCRYTOJT-UHFFFAOYSA-N | |
| 517165 | |
| azanium;acetate |
Specifications
| 631-61-8 , 7732-18-5 | |
| 61.0, 40.82 | |
| ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
| CC(=O)[O-].[NH4+] | |
| 77.083 | |
| CHEBI:62947 |
| 59.0, 39.22 | |
| C2H7NO2 | |
| USFZMSVCRYTOJT-UHFFFAOYSA-N | |
| azanium;acetate | |
| 517165 |