missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, 40% (w/w), Ricca Chemical
Supplier: Ricca Chemical Company 62016
| Quantity | 500 mL |
|---|---|
| Packaging | Natural Poly Bottle |
Chemical Identifiers
| 631-61-8 , 7732-18-5 | |
| 77.083 | |
| ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
| CHEBI:62947 | |
| CC(=O)[O-].[NH4+] |
| C2H7NO2 | |
| USFZMSVCRYTOJT-UHFFFAOYSA-N | |
| 517165 | |
| azanium;acetate |
Specifications
| 631-61-8 , 7732-18-5 | |
| 61.0, 40.82 | |
| ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
| CC(=O)[O-].[NH4+] | |
| 77.083 | |
| CHEBI:62947 | |
| 500 mL |
| 59.0, 39.22 | |
| C2H7NO2 | |
| USFZMSVCRYTOJT-UHFFFAOYSA-N | |
| azanium;acetate | |
| 517165 | |
| Natural Poly Bottle |