missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, 40% (w/w), Ricca Chemical

Supplier:  Ricca Chemical Company RI6201

Quantity 4 L
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. 6201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

631-61-8 , 7732-18-5
77.083
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CHEBI:62947
CC(=O)[O-].[NH4+]
C2H7NO2
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate

Specifications

631-61-8 , 7732-18-5
61.0, 40.82
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CC(=O)[O-].[NH4+]
77.083
CHEBI:62947
4 L
59.0, 39.22
C2H7NO2
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
Natural Poly Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions