missing translation for 'onlineSavingsMsg'
Learn More

Ammonium 1-Pyrrolidinecarbodithioate 95.0+%, TCI America™

Catalog No. P064425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P064425G 25 g
P0644250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P064425G Supplier TCI America Supplier No. P064425G

Chemical Identifiers

CAS 5108-96-3
Molecular Formula C5H8NS2-
Molecular Weight (g/mol) 146.246
MDL Number MFCD00012720
InChI Key VSWDORGPIHIGNW-UHFFFAOYSA-M
Synonym ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium
PubChem CID 4311638
IUPAC Name pyrrolidine-1-carbodithioate
SMILES C1CCN(C1)C(=S)[S-]

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 25 g
Formula Weight 164.29
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ammonium 1-Pyrrolidinecarbodithioate [Reagent for Atomic Absorption Analysis]
EINECSNumber (5)-0111
TSCA Yes
Recommended Storage Freezer