missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Standard, 1 ppm NH3, Ricca Chemical

Catalog No. 611532
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
611532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 611532 Supplier Ricca Chemical Company Supplier No. 611.532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 3.0E-4
CAS Max % 100.0, 3.0E-4
Equivalence 1 mL = 0.001 mg NH
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Ion Selective Electrode
Solubility Miscible with water
Solution Type Calibration Standard
Type Calibration Standard
Quantity 1 L
Grade Laboratory
Chemical Name or Material Ammonia Standard
Show More Show Less