missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Standard, 1000 ppm NH3 (822 ppm N), Ricca Chemical
Supplier: Ricca Chemical Company 61532
Specifications
| Ammonia Standard | |
| Natural Poly Bottle | |
| 98.0,0.31 | |
| 1mL = 1mgNH3 | |
| MFCD00011420 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 1 L | |
| CHEBI:31206 |
| 1000ppm NH3 | |
| 12125-02-9 , 7732-18-5 | |
| 100.0,0.32 | |
| ClH4N | |
| 100°C | |
| N.Cl | |
| 53.49 | |
| 25517 |
Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| CHEBI:31206 | |
| N.Cl |
| ClH4N | |
| MFCD00011420 | |
| 25517 | |
| amine hydrochloride |