missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Standard, 1000 ppm NH3 (822 ppm N), Ricca Chemical

Catalog No. p-2884480
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6154 120 mL Natural Poly Bottle
61516 500 mL Natural Poly Bottle
61532 1 L Natural Poly Bottle
6151 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6154 Supplier Ricca Chemical Company Supplier No. 6154
Only null left

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.31
CAS Max % 100.0, 0.32
Equivalence 1mL = 1mgNH3
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Ammonia Standard, 1000ppm NH3