missing translation for 'onlineSavingsMsg'
Learn More

Ammonia solution 25%, Suprapur™, MilliporeSigma™

Numéro de catalogue. p-4640337
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
M1054280250 250 mL Plastic Bottle
M1054281000 1 L Plastic Bottle
M1054282500 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. M1054280250 Fournisseur Technidata Code fournisseur 1.05428.0250
Il en reste null

Identifiants chimiques

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Spécifications

Color Colorless
Assay Percent Range 25%
Density 0.903g/cm3 (20°C)
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
Solubility Information Soluble (20°C)
Quantity 250 mL
Explosion Limit 15.4 to 33.6%(V)
Grade Suprapur
Packaging Plastic Bottle
Vapor Pressure 483 hPa (20°C)
Physical Form Liquid
Chemical Name or Material Suprapur™
Afficher plus de résultats Afficher moins de résultats
Hazard Category SignalWord: Danger
Recommended Storage Store below +25°C