missing translation for 'onlineSavingsMsg'
Learn More

Ammonia ISE Fill Solution, Fisherbrand

Catalog No. ACCU0803500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0803500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0803500 Supplier Ricca Chemical Company Supplier No. ACCU0803500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-21-1 , 7783-90-6 , 12125-02-9 , 7732-18-5
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Color Colorless
Concentration 1X
For Use With (Application) Water Analysis
For Use With (Equipment) Ion Selective Electrode
Physical Form Liquid
Quantity 500 mL
Solution Type Ionic Strength Adjustor
Odor Odorless
Packaging Natural Poly Bottle
Grade Laboratory
Chemical Name or Material Ammonia ISE Fill Solution
Show More Show Less