missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, ca. 7N solution in methanol, AcroSeal™

Catalog No. AC428388000
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC428388000 800 mL
AC428381000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428388000 Supplier Thermo Scientific Chemicals Supplier No. 428388000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7664-41-7 | H3N | 17.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Ammonia
Color Colorless to Yellow
Name Note 7N solution in methanol
CAS 7664-41-7, 67-56-1
Molecular Formula H3N
Linear Formula NH3
MDL Number MFCD00011418
Concentration 6.0 to 7.5N
Density 0.7700g/mL
Merck Index 15, 488
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILES N
IUPAC Name azane
Molecular Weight (g/mol) 17.03
PubChem CID 222
ChEBI CHEBI:16134
Formula Weight 17.03
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.77
Quantity 800 mL
Flash Point 14°C
Physical Form Liquid
Show More Show Less

RUO – Research Use Only