missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, 2M in methanol

Catalog No. AAH27080AE
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27080AE 100 mL
AAH27080AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH27080AE missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H27080AE

CAS: 7664-41-7 | H3N | 17.03 g/mol

Ammonia, 2M in methanol is used as a solvent in organic reactions and as a mobile phase in thin layer chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-41-7
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
PubChem CID 222
ChEBI CHEBI:16134
IUPAC Name azane
SMILES N

Specifications

Linear Formula NH3
UN Number UN2924
Concentration 2M in methanol
Density 0.787
Merck Index 14,492
Solubility Information Miscible with water.
Formula Weight 17.03
Quantity 100 mL
Flash Point 6°C (43°F)
Sensitivity Hygroscopic
Chemical Name or Material Ammonia
Show More Show Less
missing translation for 'healthHazard1' H225-H301+H311+H331-H315-H318-H336-H370-H372
missing translation for 'healthHazard2' GHS H Statement
H225-H311-H331-H370-H318-H315
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes serious eye damage.
Causes skin irritation.
missing translation for 'healthHazard3' P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P310-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only