missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Amlexanox

Catalog No. 485750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
485750 50 mg
485710 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 485750 Supplier R&D Systems™ Supplier No. 4857/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Selective inhibitor of TBK

Selective inhibitor of TANK-binding kinase 1 (TBK1) and IKKε (IC50 values are ∽1-2 μM). Displays no effect on IKKα or IKKβ at these concentrations. Reversibly lowers weight, increases insulin sensitivity, and reduces inflammation and steatosis in three mouse models of obesity. Exhibits antiallergic activity; inhibits the release of histamine from rat mast cells. Also binds to Hsp90 and inhibits C-terminal chaperone activity in vitro.

Chemical Identifiers

CAS 68302-57-8
Molecular Formula C16H14N2O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 298.29
Purity >99%
Molecular Weight (g/mol) 298.30
Chemical Name or Material Amlexanox
Recommended Storage Store at -20°C