missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amisulpride 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 127°C |
| Color | White |
| Quantity | 100 mg |
| Formula Weight | 369.48 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Amisulpride |
Sécurité et manipulation
| RTECSNumber | CV2308701 |
| TSCA | No |
| Recommended Storage | Refrigerator |