missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminonaphtholsulfonic Acid Solution, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 85.0, 12.71, 0.42, 0.21 |
| CAS Max % | 88.0, 13.23, 0.44, 0.22 |
| Grade | Laboratory |
| Packaging | Amber Poly Bottle |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Aminonaphtholsulfonic Acid Solution, Reducing Agent for Silica |