missing translation for 'onlineSavingsMsg'
Learn More

Aminomethanesulfonic Acid 97.0+%, TCI America™

Catalog No. A085225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A085225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A085225G Supplier TCI America Supplier No. A085225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13881-91-9
Molecular Formula CH5NO3S
Molecular Weight (g/mol) 111.115
MDL Number MFCD00008124
InChI Key OBESRABRARNZJB-UHFFFAOYSA-N
Synonym methanesulfonic acid, amino, aminomethanesulphonic acid, aminomethyl sulfonic acid, aminomethylsulfonic acid, unii-ola224z482, methanesulfonic acid, 1-amino, aminomethanesulfonicacid, sodium aminomethanesulphonate, aminomethanesulfonic, aminomethanesulfoniccid
PubChem CID 83791
IUPAC Name aminomethanesulfonic acid
SMILES C(N)S(=O)(=O)O

Specifications

Color White
Quantity 25 g
UN Number 3261
Formula Weight 111.12
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aminomethanesulfonic Acid
EINECSNumber (2)-3405
TSCA Yes