missing translation for 'onlineSavingsMsg'
Learn More

Aminomalononitrile p-toluenesulfonate, 97+%

Catalog No. AAL0900114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0900114 25 g
AAL0900106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0900114 Supplier Thermo Scientific Chemicals Supplier No. L0900114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5098-14-6 | C10H11N3O3S | 253.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5098-14-6
Molecular Formula C10H11N3O3S
Molecular Weight (g/mol) 253.276
MDL Number MFCD00012848
InChI Key MEUWQVWJLLBVQI-UHFFFAOYSA-N
Synonym aminomalononitrile p-toluenesulfonate, 2-aminomalononitrile 4-methylbenzenesulfonate, aminomalononitrile p-toluenesulphonate, aminomalononitrile p-toluenesulfonic acid, aminomalononitrile tosylate, aminomalononitrile-p-toluene sulfonate, 2-aminomalononitrile 4-methylbenzenesulphonate, 2-aminomalononitrile p-toluenesulfonate, acmc-209krb, dicyanomethanaminium 4-methylbenzenesulfonate
PubChem CID 563049
IUPAC Name 2-aminopropanedinitrile;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C(#N)C(C#N)N

Specifications

Melting Point ∽169°C (decomposition)
Quantity 25 g
UN Number UN3439
Beilstein 4050747
Formula Weight 253.28
Percent Purity ≥97%
Chemical Name or Material Aminomalononitrile p-toluenesulfonate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 225-817-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only