missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amino-Functionalized Silica Gel, Spherical, 40-75 μm

Catalog No. 111012883
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012883 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012883 Supplier MilliporeSigma™ Supelco™ Supplier No. 79297100G

Amino-functionalized silica gel spherical is monodisperse in size and shape.

Amino-functionalized silica gel spherical is prepared and characterized by thermogravimetery, X-ray diffraction, FT-IR, scanning electron microscopy (SEM) and N2 adsorption-desorption techniques.

Specifications

Particle Size 40 to 75 μm
Pore Size 110 Å
Grade High-purity Grade
Quantity 100 g
Surface Area (Metric) 500 m2/g (approx.)
Description Nitrogen content, >1.0%
Physical Form Powder
CAS 126850-14-4