missing translation for 'onlineSavingsMsg'
Learn More

Amine-Sulfuric Acid Reagent, For Sulfide, Certified, LabChem™

Catalog No. LC108801
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC108801 500 mL
LC108802 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC108801 Supplier LabChem Supplier No. LC108801
Only null left

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5 , 62778-12-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Amber to Pink
CAS Max % 58.91, 40.41, 0.68
Linear Formula C18H26N4O4
UN Number UN1830
Density 1.49g/mL
Solubility Information Soluble in water
Formula Weight 181.21
Grade Certified
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.49g/mL
Suitability Passes Test
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors; Nitrogen oxides
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Amine-Sulfuric Acid Reagent, For Sulfide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature