missing translation for 'onlineSavingsMsg'
Learn More

Amiloride Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013352
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013352 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013352 Supplier Merck Emd Millipore Supplier No. PHR18391G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 17440-83-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C6H8 ClN7 O · HCl · 2H2 O
Synonym Amiloride hydrochloride dihydrate; N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 302.12
Grade Certified Reference Material