missing translation for 'onlineSavingsMsg'
Learn More

Amido Black 10B

Catalog No. AAA1137422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1137422 100 g
AAA1137414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1137422 Supplier Thermo Scientific Chemicals Supplier No. A1137422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as a histological, protein, and nucleic acid dye used in electrophoresis. | CAS: 1064-48-8 | C22H14N6Na2O9S2 | 616.487 g/mol

Amido Black 10B is used as a histological, protein, and nucleic acid dye used in electrophoresis. Naphthol Blue Black is an important and commonly used industrial dye. This dye is also used in rapid staining of protein bands on nitrocellulose membranes such as Western blots. Naphthol Blue Black can be utilized as a nucleic acid stain in SDS-Page gels. Additionally this compound has been untilized in cell proliferation assays and histochemical studies. It is also used in criminal investigations to detect blood present with latent fingerprints. They are also used in paints, inks, plastics and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1064-48-8
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.487
MDL Number MFCD00004017
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
Synonym C.I. 20470,Naphthalene Black 10B
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Packaging Glass bottle
Sensitivity Hygroscopic
Beilstein 5374263
Solubility Information It is soluble in water (10mg/ml at 25°C),and ethanol.
Quantity 100 g
Physical Form Powder
Formula Weight 616.49
Chemical Name or Material Amido Black 10B
Hazard Category H315-H317-H319-H335-H372-H500
Precautionary Statement P261-P264b-P270-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P314-P332+P313-P362-P501c
EINECSNumber 213-903-1
RTECSNumber QJ6196000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only