missing translation for 'onlineSavingsMsg'
Learn More

Amido Black 10B, Spectrum™ Chemical

Catalog No. 18608566
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608566 25 g Amber Glass Bottle
18608565 100 g Amber Glass Bottle
18608564 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608566 Supplier Spectrum Chemical Mfg Cor Supplier No. AM11025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1064-48-8 | 616.52 g/mol

Chemical Identifiers

CAS 1064-48-8

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Color Dark Brown
Quantity 25 g
Molecular Weight (g/mol) 616.52
Grade Reagent
Chemical Name or Material Amido Black 10B

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.