missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amido Black 10B, Electrophoresis Reagent

Catalog No. AAJ6679822
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ6679822 100 g Glass bottle
AAJ6679814 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6679822 Supplier Thermo Scientific Chemicals Supplier No. J6679822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1064-48-8 | C22H14N6Na2O9S2 | 616.487 g/mol

Used to stain proteins in polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1064-48-8
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.487
MDL Number MFCD00004017
InChI Key HKBVRFLHNUEVRO-UHFFFAOYSA-L
Synonym C.I. 20470,Naphthalene Black 10B
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Packaging Glass bottle
Sensitivity Hygroscopic
Beilstein 5374263
Quantity 100 g
Physical Form Powder
Formula Weight 616.49
Chemical Name or Material Amido Black 10B
Hazard Category H315-H317-H319-H335-H372-H500
Precautionary Statement P261-P264b-P270-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P314-P332+P313-P362-P501c
EINECSNumber 213-903-1
RTECSNumber QJ6196000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only