missing translation for 'onlineSavingsMsg'
Learn More

Amezinium Methyl Sulfate 98.0+%, TCI America™

Catalog No. A24255G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24255G 5 g
A24251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24255G Supplier TCI America Supplier No. A24255G
Only null left

Chemical Identifiers

CAS 30578-37-1
Molecular Formula C12H15N3O5S
Molecular Weight (g/mol) 313.33
MDL Number MFCD00865972
InChI Key ZEASXVYVFFXULL-UHFFFAOYSA-N
Synonym 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulfate
PubChem CID 71926
IUPAC Name 4-amino-6-methoxy-1-phenyl-1λ⁵-pyridazin-1-ylium methyl sulfate
SMILES COS([O-])(=O)=O.COC1=CC(N)=CN=[N+]1C1=CC=CC=C1

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 5 g
Formula Weight 313.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Amezinium Methyl Sulfate
RTECSNumber UR6066500
TSCA No