Learn More
Cytiva Amersham™ Streptavidin-HRP Conjugate
Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications
Supplier: Cytiva RPN1231100UL
Description
- High specificity reagents
- Conjugation achieved using the periodate coupling method described by Nakane and co-workers
- Sold on the basis of titer in an ELISA and performance in a western blot
Specifications
| Suitable for use in ECL™, western blotting ,ELISA, immunocytochemistry | |
| 2mL of Phosphate Buffered Saline (0.1M Sodium Phosphate, 0.1M Sodium Chloride, pH 7.4) containing 1%(w/v) Bovine Serum Albumin and an antimicrobial agent | |
| 100 mL |