Learn More
AME-CLAMP PINCH CLAMP 10/PK
Numéro de catalogue. 501109801
Encompass
WL GORE Clamp,Ame Clamp, WL GORE/Amesil Clamp,uniquely designed closure for tubing or hose that requires Higher clamp pressures. Can be utilized with a variety of tubing and hose sizes. Quckly installed with a positive swingaway locking screw. Sanitary , Reusable, Lightweight , Heavy duty high strength polymer construction, PK/10 ea
Numéro de catalogue. 501109801
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
WL GORE Clamp,Ame Clamp, WL GORE/Amesil Clamp,uniquely designed closure for tubing or hose that requires Higher clamp pressures. Can be utilized with a variety of tubing and hose sizes. Quckly installed with a positive swingaway locking screw. Sanitary , Reusable, Lightweight , Heavy duty high strength polymer construction, PK/10 ea