missing translation for 'onlineSavingsMsg'
Learn More

Ambroxol hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013644
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013644 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013644 Supplier Merck Emd Millipore Supplier No. A97975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ambroxol hydrochloride is a mucolytic expectorant and a metabolite of bromhexine. It is used in the treatment of respiratory disorders characterized by viscous or excessive mucus.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 23828-92-4
Physical Form Neat
Linear Formula C13H18 Br2N2 O · HCl
MDL Number MFCD00078932
Synonym 2-Amino-3,5-dibromo-N-(trans-4-hydroxycyclohexyl)benzylamine
Type Analytical Standard
Quantity 5 g
Formula Weight 414.56
Grade Analytical Standard