missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vortex Adapter-60

Catalog No. am10014
Encompass
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM10014 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM10014 Supplier Invitrogen™ Supplier No. AM10014
Only null left

The adapter's unique shaking motion promotes rapid and thorough tissue digestion. Recommended for use with MELT™ Total RNA Isolation Kit (Cat. No. AM1983).

The adapter's unique shaking motion promotes rapid and thorough tissue digestion. Recommended for use with MELT™ Total RNA Isolation Kit (Cat. No. AM1983).

  • Vortex Adapter-60 consists of a 6 in. (152 mm) platform and two different Microplate Foam Inserts
  • Each insert fits snugly into platform to accommodate either 60 microtubes or one standard microplate
  • Compatible with Vortex-Genie™ 2, Vortex-Genie 2T, and VWR™ Mini Vortex Mixers

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) Vortex
Shipping Condition Room Temperature
Product Line Ambion
Quantity 1 each
Type Tools and Accessories

For Research Use Only. Not for use in diagnostic procedures.