missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T7 RNA Polymerase-Plus™ Enzyme Mix (200 U/μL)

Catalog No. AM2718
Encompass
Change view
Click to view available options
Quantity:
30000 units, 200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM2718 30000 units, 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM2718 Supplier Invitrogen™ Supplier No. AM2718

missing translation for 'validMainframeMsg'

Includes

One tube containing 30,000 units at a concentration of 20U/μL

High-purity enzyme that's a mix of cloned RNA polymerase and RNase inhibitor

 

  • This enzyme has a high specificity for its promoter, transcribing large amounts of RNA with no cross talk between promoters
  • It is supplied in one tube containing 30,000U at a concentration of 200U/μL
  • The enzyme incorporates radiolabeled nucleotides along with many modified nucleotides
  • Inclusion of RNase inhibitor provides broad-spectrum protection against enzymatic degradation of the synthesized RNA, making the enzyme ideal for the synthesis of RNA for blot and in situ hybridization probes, RPA analysis, in vitro translation, and antisense RNA 

Unit Definition: One unit is the amount of T7 RNA polymeras T7 RNA Polymerase-Plus™ Enzyme Mix required to catalyze the incorporation of 1 nmol of nucleoside triphosphate in 60 minutes at 37°C.

DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, in vitro Transcription, Nucleic Acid Labeling & Oligo Synthesis, RNA Labeling

Order Info

Shipping Condition: Dry ice

Specifications

Content And Storage Store at –20°C.
Shipping Condition Dry Ice
Polymerase T7 RNA Polymerase
Product Line Ambion
Promoter T7
Quantity 30000 units, 200 μL
Concentration 200 U/μL
Source DNA
Can I substitute the enzyme mix in the MEGAscript T7 Transcription Kit (Cat. No. AM1333) with T7 RNA Polymerase-Plus Enzyme Mix (Cat. No. AM2718)?

No, we do not recommend substituting the enzyme mix in the MEGAscript T7 Transcription Kit (Cat. No. AM1333) with T7 RNA Polymerase-Plus Enzyme Mix (Cat. No. AM2718).

For Research Use Only. Not for use in diagnostic procedures.